Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rp101988 | 2990018-57-8 | 98.8% | C23H22N4O4 | 10MG
SDP

Catalog No. 5000261487
Encompass_Preferred

RP101988 is the major active metabolite of ozanimod and a selective, potent sphingosine-1-phosphate receptor 1 (S1PR1) agonist used for in vitro and pharmacological research. The compound is characterized for potency, purity, solubility, and storage stability to support assay development and preclinical studies.

  • Selective S1PR1 agonist with EC50 0.19 nM (S1PR1) and 32.8 nM (S1PR5).
  • High purity suitable for research use (98.8%).
  • Available as solid and as a 10 mM DMSO solution for experimental flexibility.
  • Soluble in DMSO at 125 mg/mL; ultrasonic assistance recommended.
  • Recommended storage conditions for powder and solutions to preserve stability.

Catalog No. 50-002-61487 Supplier Medchemexpress LLC Supplier No. HY13657810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

RP101988 is the major active metabolite of ozanimod and a selective, potent sphingosine-1-phosphate receptor 1 (S1PR1) agonist used for in vitro and pharmacological research. The compound is characterized for potency, purity, solubility, and storage stability to support assay development and preclinical studies.

  • Selective S1PR1 agonist with EC50 0.19 nM (S1PR1) and 32.8 nM (S1PR5).
  • High purity suitable for research use (98.8%).
  • Available as solid and as a 10 mM DMSO solution for experimental flexibility.
  • Soluble in DMSO at 125 mg/mL; ultrasonic assistance recommended.
  • Recommended storage conditions for powder and solutions to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.