Learn More
Waters Corp Waters XBridge BEH Shield RP18 VanGuard Cartridge, 130Å, 5 μm
Catalog No. NC1486138 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Shield RP18 VanGuard Cartridges extend analytical column lifetime and performance by filtering particulate contamination. They are optimized for 4.6 mm I.D. analytical columns with 5 μm sorbent particles, ensuring precise and reliable separations in LC, bioanalysis, pharmaceutical, and environmental analysis.
  • Extends analytical column lifetime and performance.
  • Optimized for 4.6 mm I.D. analytical columns with 5 μm sorbent particles.
  • Constructed from styrene-divinylbenzene copolymer.
  • Effectively filters out unwanted particles.
  • Ensures optimal purification and reduces contamination from mobile phases with a pH range of 2 to 8.
  • Easy to use and install.
  • Maintains column integrity and enhances chromatographic separation quality.
Catalog No. NC1486138 Supplier Waters Corp Supplier No. 186007807
XBridge BEH Shield RP18 VanGuard Cartridges extend analytical column lifetime and performance by filtering particulate contamination. They are optimized for 4.6 mm I.D. analytical columns with 5 μm sorbent particles, ensuring precise and reliable separations in LC, bioanalysis, pharmaceutical, and environmental analysis.
  • Extends analytical column lifetime and performance.
  • Optimized for 4.6 mm I.D. analytical columns with 5 μm sorbent particles.
  • Constructed from styrene-divinylbenzene copolymer.
  • Effectively filters out unwanted particles.
  • Ensures optimal purification and reduces contamination from mobile phases with a pH range of 2 to 8.
  • Easy to use and install.
  • Maintains column integrity and enhances chromatographic separation quality.