Learn More
Aviva Systems Biology Corp RP2 Antibody - middle region (ARP56724_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56724_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RP2. Alternate names: KIAA0215, TBCCD2, XRP2, NME10, DELXp11.3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61964