Learn More
Pyramex™ RP2001 Series Reusable Ear Plugs
Catalog No. 19155495 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
26 dB
Catalog No. Noise Reduction Rating (NRR)
19-155-495 26 dB
1 options
Catalog No. 19-155-495 Supplier Pyramex™ Supplier No. RP2001
Add to Cart
Add to Cart
Reusable RP2001 Flange-style flexible rubber form a cozy seal for all size ear canals.
  • Triple flange reuseable TPR Rubber ear plugs
  • Plug has a pocket of air that makes it lightweight
  • Packaged in a convenient dispenser box – 100 corded pair per box
  • Material is not known to cause adverse affect to user hygiene or health, nor is likely to cause irritation, during normal use
  • Latex-free
Specifications
ANSI S3.1974 Yes
Cord Color Yellow
Cord Material PVC
Size Universal
For Use With (Application) Assembly, Construction, Manufacturing, Transportation, Utilities, and Other Various Industries
High Visibility Yes
Packaging Type Dispenser box
Pairs per Package 100 Pairs/Pk.
Plug Material Thermoplastic rubber
Plug Color Blue
Noise Reduction Rating (NRR) 26 dB
Corded Corded
Plug Shape Flanged
Product Type Reusable Earplugs
Disposable Reusable
Metal Detectable No
Dielectric No