Learn More
Aviva Systems Biology Corp RPA1 Antibody - middle region (ARP56526_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56526_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RPA1. Alternate names: HSSB, REPA1, RF-A, RP-A, RPA70, MST075ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-61893