Learn More
Aviva Systems Biology Corp RPAIN Antibody - C-terminal region (ARP41887_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41887_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPAIN. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human RPAIN. Alternate names: FLJ25625, FLJ30490, FLJ42429, HRIP, MGC4189, RIPApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-284-963