Learn More
Cayman Chemical R-Palmitoyl-(1-methyl) Ethanolamide, Purity: ≥98%, CAS: 142128-47-0, 50 mg
SDP
Catalog No. 502892913
Encompass_Preferred
A synthetic analog of PEA which incorporates an (R)-methyl group vicinal to the alcohol on the ethanolamine moiety, which may protect the molecule from enzymatic hydrolysis, leading to prolonged duration of action and enhanced potency in vivo; expected to show enhancement of the inhibitory activity of PEA on mast cells and other cells known to express the CB2 receptor
Catalog No. 50-289-2913 Supplier Cayman Chemical Supplier No. 9035350 MG
A synthetic analog of PEA which incorporates an (R)-methyl group vicinal to the alcohol on the ethanolamine moiety, which may protect the molecule from enzymatic hydrolysis, leading to prolonged duration of action and enhanced potency in vivo; expected to show enhancement of the inhibitory activity of PEA on mast cells and other cells known to express the CB2 receptor