Learn More
Braintree Scientific RPC-1-Rat PieCage to 250g,41cm Dia 6.75cm H-Aerosol Delivery
SDP
Catalog No. 501955745
Encompass
Rat Pie Cages - easy to clean and to use. Circular cages hold up to 11 rats for whole body irradiation. Notched removable lid can be dialed to any of the ventilated pie-shaped chambers. Also use for temporary transport or storage of animals for acute studies. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5745 Supplier Braintree Scientific Supplier No. RPC1AERO
Rat Pie Cages - easy to clean and to use. Circular cages hold up to 11 rats for whole body irradiation. Notched removable lid can be dialed to any of the ventilated pie-shaped chambers. Also use for temporary transport or storage of animals for acute studies. **Sold for research purposes only and not for human use**