Learn More
Braintree Scientific RPC-2 Rat Pie Cage Custom,46cm Dia 8.cm H
SDP
Catalog No. 501955749
Encompass
Rat Pie Cages - easy to clean and to use. Circular cages hold up to 11 rats for whole body irradiation. Notched removable lid can be dialed to any of the ventilated pie-shaped chambers. Also use for temporary transport or storage of animals for acute studies. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5749 Supplier Braintree Scientific Supplier No. RPC2STD
Rat Pie Cages - easy to clean and to use. Circular cages hold up to 11 rats for whole body irradiation. Notched removable lid can be dialed to any of the ventilated pie-shaped chambers. Also use for temporary transport or storage of animals for acute studies. **Sold for research purposes only and not for human use**