Learn More
Aat Bioquest RPE-Cy5-streptavidin conjugate (100 ug)
SDP
Catalog No. 502249626
Encompass_Preferred
Streptavidin conjugates are widely used together with a biotin conjugate for specific detection of a variety of biological molecules since streptavidin has a very high binding affinity for biotin and its conjugates. This catalog is for 100 ug.
This item is not returnable. View return policy
Catalog No. 50-224-9626 Supplier Aat Bioquest Supplier No. 16918
Streptavidin conjugates are widely used together with a biotin conjugate for specific detection of a variety of biological molecules since streptavidin has a very high binding affinity for biotin and its conjugates. This catalog is for 100 ug.