Learn More
Aviva Systems Biology Corp RPESP Antibody - middle region (ARP53142_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53142_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPESP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RPESP. Alternate names: FLJ40021, RPESPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60942