Learn More
Aviva Systems Biology Corp Rph3a Antibody - N-terminal region (ARP59498_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59498_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rph3a. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2900002P20Rik, AU022689, AW108370ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-721