Learn More
TARGETMOL CHEMICALS INC RPI-1 25MG
SDP
Catalog No. 502580918
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. RPI-1 is a competitive potent ATP-dependent Ret kinase inhibitor. purity: 98%
Catalog No. 50-258-0918 Supplier TARGETMOL CHEMICALS INC Supplier No. T228225MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. RPI-1 is a competitive potent ATP-dependent Ret kinase inhibitor. purity: 98%