Learn More
Research Products International Corp RPI FK-506 [Tacrolimus] [Fujimycin], 100 Milligrams

Supplier: Research Products International Corp F221000.1
Substance Name: [Tacrolimus] [Fujimycin]
CAS Number: 104987-11-3
Molecular Formula: C44H69NO12
Molecular Weight: 804.0
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.