Learn More
Research Products International Corp RPI Color Coded Multi-Purpose Laboratory Tape, 1 Inch Core, 3/4 Inch Wide, 500 Inches per Roll, Violet
SDP
Catalog No. 50550386
Encompass
Laboratory tape adheres to any clean dry surface including glass, plastic, metal, paper, or rubber and removes without leaving a sticky residue. Can be marked with pencil, pen or solvent ink markers. Tape adhesive is oilproof, waterproof and acid resistant.
Catalog No. 50-550-386 Supplier Research Products International Corp Supplier No. 563400V
Laboratory tape adheres to any clean dry surface including glass, plastic, metal, paper, or rubber and removes without leaving a sticky residue. Can be marked with pencil, pen or solvent ink markers. Tape adhesive is oilproof, waterproof and acid resistant.