Learn More
Research Products International Corp RPI PIPES [Piperazine-N-N'-bis(2-ethanesulfonic acid)], 5 Kilograms
SDP
Catalog No. 501976858
Encompass_Preferred
Heavy metals: <10 ppm
pka (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5
Catalog No. 50-197-6858 Supplier Research Products International Corp Supplier No. P401405000.0
Heavy metals: <10 ppm
pka (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5