Learn More
Research Products International Corp RPI Pipette Filler, Rechargeable, Motorized
SDP
Catalog No. 50550295
Encompass
The RPI Pipette Filler fits all glass or plastic pipettes from 1 - 100 ml. Features eight speeds for aspirating and dispensing liquids with high accuracy. Lightweight ergonomic design allows one hand operation and reduces pipetting fatigue. Individual controls are provided for aspirate and dispense operations. Fills a 50 ml pipette in less than five seconds and dispenses liquid by gravity flow or motorized operation. Built-in Lithium-ion battery provides a long run time between charges. LCD panel displays speed selection and battery charge. Supplied with two hydrophobic 0.45 um filters, charger and a one year warranty.
Catalog No. 50-550-295 Supplier Research Products International Corp Supplier No. 248646
The RPI Pipette Filler fits all glass or plastic pipettes from 1 - 100 ml. Features eight speeds for aspirating and dispensing liquids with high accuracy. Lightweight ergonomic design allows one hand operation and reduces pipetting fatigue. Individual controls are provided for aspirate and dispense operations. Fills a 50 ml pipette in less than five seconds and dispenses liquid by gravity flow or motorized operation. Built-in Lithium-ion battery provides a long run time between charges. LCD panel displays speed selection and battery charge. Supplied with two hydrophobic 0.45 um filters, charger and a one year warranty.