Learn More
Research Products International Corp RPI Vortex Adapter for 1.5 ml Micro-Centrifuge Tube, 20 Tube Capacity
SDP
Catalog No. 501977786
Encompass
Securely holds 1.5 ml micro centrifuge tubes in an 8 or 20 capacity disk for simultaneous vortexing. Tube contents are mixed by holding the shaft against the rubber pad of a vortexer. It is made of polypropylene with a 16.5 cm (6 1/2 inch) long shaft and 2.54 cm (1 inch) diameter ball handle.
Catalog No. 50-197-7786 Supplier Research Products International Corp Supplier No. F370800020
Securely holds 1.5 ml micro centrifuge tubes in an 8 or 20 capacity disk for simultaneous vortexing. Tube contents are mixed by holding the shaft against the rubber pad of a vortexer. It is made of polypropylene with a 16.5 cm (6 1/2 inch) long shaft and 2.54 cm (1 inch) diameter ball handle.