Learn More
Martor #5232 Allfit Trapezoid Blade (100 Per Box)
Supplier:  Martor 5232.70
Encompass_Preferred
ALLFIT trapezoid blade, deep-edgedLength: 53.00 mmHeight: 19.00 mmThickness: 0.63 mmPackaging: 100 per box
Catalog No. NC9407270