Learn More
Martor Allfit Trapezoid Blade (100 Per Box)
Supplier:  Martor 99.70
Encompass_Preferred
ALLFIT Trapezoid Blade, Deep EdgedLength: 60.00 mmHeight: 19.00 mmThickness: 0.63 mmPackaging: 100 per box
Catalog No. NC9944826