Learn More
MSC Goodway Thermofiber Cloths, Non-Abrasive, 5-Pack, Washable
Catalog No. 502837402
Encompass
These non-abrasive thermofiber cloths are designed for use with vapor steam cleaners. They are highly absorbent, cleaning effectively without leaving fibers, and can pick up dirt, grime, grease, and bacteria.
  • Thicker than microfiber material
  • Absorbs up to 7 times its own weight
  • Machine washable 500-700 times
  • Fits on triangular and rectangular brushes
  • Comes in a pack of 5
Catalog No. 50-283-7402 Supplier MSC Supplier No. 56547789
These non-abrasive thermofiber cloths are designed for use with vapor steam cleaners. They are highly absorbent, cleaning effectively without leaving fibers, and can pick up dirt, grime, grease, and bacteria.
  • Thicker than microfiber material
  • Absorbs up to 7 times its own weight
  • Machine washable 500-700 times
  • Fits on triangular and rectangular brushes
  • Comes in a pack of 5