Learn More
MSC RPL TIP F/SKULL SCREW
Catalog No. 5000677234
Encompass
These replacement tips are designed for skull screw earplugs, offering effective hearing protection. They provide 30dB of noise reduction and are suitable for repeated use.
  • Gray color
  • Pack of 10 pairs
  • Reusable design
  • Noise reduction of 30dB
Catalog No. 50-006-77234 Supplier MSC Supplier No. 32480196
These replacement tips are designed for skull screw earplugs, offering effective hearing protection. They provide 30dB of noise reduction and are suitable for repeated use.
  • Gray color
  • Pack of 10 pairs
  • Reusable design
  • Noise reduction of 30dB