Learn More
MSC RPL VISOR F/9100FX
Catalog No. 5000792764
Encompass
These clear polycarbonate replacement visors are designed for front-mounted applications. Each visor measures 8 in. wide by 4 in. high and is 0.06 in. thick, offering reliable protection.
  • Made from durable polycarbonate material
  • Clear lens provides unobstructed view
  • Front mount type for easy installation
  • Window height is 4 in.
  • Window width is 8 in.
  • Window thickness is 0.06 in.
  • Meets ANSI Z87.1-2010 and CSA Z94.3 standards for safety
  • Compatible with 9100 MP, 9100 FX, Speedglas 9100 FX, and 9100 FX-Air series welding helmets
  • Supplied in a convenient 5-pack
Catalog No. 50-007-92764 Supplier MSC Supplier No. 58427485
These clear polycarbonate replacement visors are designed for front-mounted applications. Each visor measures 8 in. wide by 4 in. high and is 0.06 in. thick, offering reliable protection.
  • Made from durable polycarbonate material
  • Clear lens provides unobstructed view
  • Front mount type for easy installation
  • Window height is 4 in.
  • Window width is 8 in.
  • Window thickness is 0.06 in.
  • Meets ANSI Z87.1-2010 and CSA Z94.3 standards for safety
  • Compatible with 9100 MP, 9100 FX, Speedglas 9100 FX, and 9100 FX-Air series welding helmets
  • Supplied in a convenient 5-pack