Learn More
Aviva Systems Biology Corp RPL10 Antibody - C-terminal region (ARP64376_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64376_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL10. It was validated on Western Blot. . Alternate names: DKFZp686J1851, DXS648, DXS648E, FLJ23544, FLJ27072, NOV, QMApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-288-249