Learn More
Aviva Systems Biology Corp RPL11 Antibody - C-terminal region (ARP64308_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64308_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL11. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human RPL11. Alternate names: DBA7, GIG34ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-233