Learn More
Proteintech Group Inc RPL11 Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 162771AP150UL
Encompass_Preferred
The RPL11 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RPL11. This antibody recognizes human, mouse, rat antigen. The RPL11 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Catalog No. 50-225-6408