Learn More
Aviva Systems Biology Corp RPL13 Antibody - C-terminal region (ARP40218_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40218_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RPL13. Alternate names: BBC1, D16S444E, MGC71373, L13, D16S44EApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57348