Learn More
Aviva Systems Biology Corp RPL13A Antibody - middle region (ARP58522_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58522_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL13A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RPL13A. Alternate names: L13A, TSTA1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-62341