Learn More
Aviva Systems Biology Corp RPL14 Antibody - C-terminal region (ARP61692_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61692_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL14. It was validated on Western Blot. . Alternate names: CAG-ISL-7, CTG-B33, L14, MGC88594, RL14, hRL14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-439