Learn More
Aviva Systems Biology Corp RPL18 Antibody - N-terminal region (ARP56127_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56127_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL18. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RPL18. Alternate names: L18ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61730