Learn More
Aviva Systems Biology Corp RPL21 Antibody - middle region (ARP65195_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65195_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL21. It was validated on Western Blot. . Alternate names: DKFZp686C06101, FLJ27458, L21, MGC104274, MGC104275, MGC71252ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-453