Learn More
Aviva Systems Biology Corp Rpl26 Antibody - middle region (ARP61600_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61600_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rpl26. It was validated on Western Blot. . Alternate names: SIG-20ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-418