Learn More
Aviva Systems Biology Corp RPL27 Antibody - middle region (ARP56132_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56132_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL27. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RPL27. Alternate names: L27ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61731