Learn More
Aviva Systems Biology Corp RPL36 Antibody - N-terminal region (ARP64986_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64986_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL36. It was validated on Western Blot. . Alternate names: DKFZp566B023, L36ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-407