Learn More
Aviva Systems Biology Corp RPL37A Antibody - middle region (ARP56137_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56137_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL37A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RPL37A. Alternate names: MGC74786, L37AApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61734