Learn More
Aviva Systems Biology Corp RPL39 Antibody - N-terminal region (ARP62105_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62105_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL39. It was validated on Western Blot. . Alternate names: L39ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-569