Learn More
Aviva Systems Biology Corp RPL5 Antibody - N-terminal region (ARP56125_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56125_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RPL5. Alternate names: MGC117339, MSTP030, L5, DBA6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61729