Learn More
Aviva Systems Biology Corp RPL6 Antibody - N-terminal region (ARP40213_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40213_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RPL6. Alternate names: L6, TXREB1, SHUJUN-2, TAXREB107ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-57344