Learn More
Aviva Systems Biology Corp RPL7A Antibody - C-terminal region (ARP61726_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61726_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL7A. It was validated on Western Blot. . Alternate names: SURF3, TRUP, L7AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-452