Learn More
ABclonal Technology RPL7A Rabbit pAb
SDP
Supplier:  ABclonal Technology A140601000UL
Encompass_Preferred
This gene encodes a ribosomal protein that is a component of the 60S subunit and belongs to the L7AE family of ribosomal proteins. It can interact with nuclear hormone receptors, such as the thyroid hormone receptor, and inhibit their ability to transactivate by preventing binding to DNA response elements. The gene is part of the surfeit gene cluster and is co-transcribed with small nucleolar RNA genes U24, U36a, U36b, and U36c located in its introns. The gene can rearrange with the trk proto-oncogene to form the chimeric oncogene trk-2h, which encodes an oncoprotein consisting of ribosomal protein L7a’s N terminus fused to the receptor tyrosine kinase domain of trk. Multiple processed pseudogenes of this gene are dispersed throughout the genome.
Catalog No. 50-001-08601