Learn More
Aviva Systems Biology Corp RPL8 Antibody - C-terminal region (ARP40216_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40216_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL8. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human RPL8. Alternate names: L8ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57346