Learn More
Aviva Systems Biology Corp RPL9 Antibody - C-terminal region (ARP40212_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40212_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RPL9. Alternate names: L9, NPC-A-16ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57343