Learn More
Electron Microscopy Sciences Dumont WA1 Tweezer Handle with Style 10A ESD Tips, 0.6 mm Tip width, 0.5 mm Tip thickness, 125 mm OAL
SDP
Catalog No. 501927499
Encompass_Preferred
These tweezers have an anti-magnetic WA1 handle with replaceable 10A-ESD tips. Tips attach with screws. Length is 125 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Black ESD
Tip width: 0.6 mm
Tip thickness: 0.5 mm
OAL: 125 mm
Catalog No. 50-192-7499 Supplier Electron Microscopy Sciences Supplier No. 10004E10ASM
These tweezers have an anti-magnetic WA1 handle with replaceable 10A-ESD tips. Tips attach with screws. Length is 125 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Black ESD
Tip width: 0.6 mm
Tip thickness: 0.5 mm
OAL: 125 mm