Learn More
Electron Microscopy Sciences Dumont WA1 Tweezer Handle with Style 17 STD Tips, 1.5 mm Tip width, 0.35 mm Tip thickness, 130 mm OAL
SDP
Catalog No. 501927531
Encompass_Preferred
These tweezers have an anti-magnetic WA1 handle with replaceable 17-STD tips. Tips attach with screws. Length is 130 mm. Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Tips: Grey STD
Tip width: 1.5 mm
Tip thickness: 0.35 mm
OAL: 130 mm
Catalog No. 50-192-7531 Supplier Electron Microscopy Sciences Supplier No. 10004S17SM
These tweezers have an anti-magnetic WA1 handle with replaceable 17-STD tips. Tips attach with screws. Length is 130 mm. Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Tips: Grey STD
Tip width: 1.5 mm
Tip thickness: 0.35 mm
OAL: 130 mm