Learn More
Electron Microscopy Sciences Dumont WA1 Tweezer Handle with Style 17B ESD Tips, 1.5 mm Tip width, 0.25 mm Tip thickness, 130 mm OAL
SDP
Catalog No. 501931623
Encompass_Preferred
These tweezers have an anti-magnetic WA1 handle with replaceable 17B-DELRIN tips. Tips attach with screws. Length is 130 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Black ESD
Tip width: 1.5 mm
Tip thickness: 0.25 mm
OAL: 130 mm
Catalog No. 50-193-1623 Supplier Electron Microscopy Sciences Supplier No. 10004E17BSM
These tweezers have an anti-magnetic WA1 handle with replaceable 17B-DELRIN tips. Tips attach with screws. Length is 130 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Black ESD
Tip width: 1.5 mm
Tip thickness: 0.25 mm
OAL: 130 mm