Learn More
Electron Microscopy Sciences Dumont WA1 Tweezer Handle with Style 20 ESD Tips, 2.5 mm Tip width, 0.5 mm Tip thickness, 1.50 mm Tip C
SDP
Catalog No. 501927503
Encompass_Preferred
These tweezers have an anti-magnetic WA1 handle with replaceable 20-ESD tips. Tips attach with screws. Length is 125 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Black ESD
Tip width: 2.5 mm
Tip thickness: 0.5 mm
Tip C: 1.50 mm
Catalog No. 50-192-7503 Supplier Electron Microscopy Sciences Supplier No. 10004E20SM
These tweezers have an anti-magnetic WA1 handle with replaceable 20-ESD tips. Tips attach with screws. Length is 125 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Black ESD
Tip width: 2.5 mm
Tip thickness: 0.5 mm
Tip C: 1.50 mm