Learn More
Electron Microscopy Sciences Dumont Style 22 Black ESD Replaceable Tweezer Tips, 0.5 mm Tip width, 1 mm Tip thickness
SDP
Catalog No. 501927505
Encompass_Preferred
These tweezers have an anti-magnetic WA1 handle with replaceable 22-ESD tips. Tips attach with screws. Length is 125 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Black ESD
Tip width: 0.5 mm
Tip thickness: 1 mm
Tip C: 2.50 mm
Catalog No. 50-192-7505 Supplier Electron Microscopy Sciences Supplier No. 10004E22SM
These tweezers have an anti-magnetic WA1 handle with replaceable 22-ESD tips. Tips attach with screws. Length is 125 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: Black ESD
Tip width: 0.5 mm
Tip thickness: 1 mm
Tip C: 2.50 mm