Learn More
Electron Microscopy Sciences Dumont WA1 Tweezer Handle with Style 3 STD Tips, 2.3 mm Tip width, Tip thickness, 125 mm OAL
SDP
Catalog No. 501927529
Encompass_Preferred
These tweezers have an anti-magnetic WA1 handle with replaceable 3-STD tips. Tips attach with screws. Length is 125 mm. Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Tips: Grey STD
Tip width: 2.3 mm
OAL: 125 mm
Catalog No. 50-192-7529 Supplier Electron Microscopy Sciences Supplier No. 10004S3SM
These tweezers have an anti-magnetic WA1 handle with replaceable 3-STD tips. Tips attach with screws. Length is 125 mm. Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Tips: Grey STD
Tip width: 2.3 mm
OAL: 125 mm