Learn More
Electron Microscopy Sciences Dumont WA1 Tweezer Handle with Style 4 Delrin Tips, 0.6 mm Tip width, 0.3 mm Tip thickness, 130 mm OAL
SDP
Catalog No. 501927515
Encompass_Preferred
These tweezers have an anti-magnetic WA1 handle with replaceable 4-ESD tips. Tips attach with screws. Length is 130 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: White Delrin
Tip width: 0.6 mm
Tip thickness: 0.3 mm
OAL: 130 mm
Catalog No. 50-192-7515 Supplier Electron Microscopy Sciences Supplier No. 10004D4SM
These tweezers have an anti-magnetic WA1 handle with replaceable 4-ESD tips. Tips attach with screws. Length is 130 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tips: White Delrin
Tip width: 0.6 mm
Tip thickness: 0.3 mm
OAL: 130 mm