Learn More
Electron Microscopy Sciences Dumont WA1 Tweezer Handle with Style 50 ESD Tips, 10 mm Tip width, 8 mm Tip thickness, 115 mm OAL
SDP
Catalog No. 501927507
Encompass_Preferred
These tweezers have an anti-magnetic WA1 handle with replaceable 50-ESD tips. Tips attach with screws. Length is 115 mm. Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Tips: Black ESD
Tip width: 10 mm
Tip thickness: 8 mm
OAL: 115 mm
Catalog No. 50-192-7507 Supplier Electron Microscopy Sciences Supplier No. 10004E50SM
These tweezers have an anti-magnetic WA1 handle with replaceable 50-ESD tips. Tips attach with screws. Length is 115 mm. Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Tips: Black ESD
Tip width: 10 mm
Tip thickness: 8 mm
OAL: 115 mm